(4S)-2,2-Dioxido-4- isopropyl -1,2,3-oxathiazolidine, N-BOC protected
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
(S)-tert-Butyl 4-isopropyl-1,2,3-oxathiazolidine-3-carboxylate 2,2-dioxide
OR400335-1G
OR400335-250MG
OR400335-5G
APOSOR400335-1GEA
212
EUR
APOSOR400335-1G
APOSOR400335-250MG
APOSOR400335-5G
(4S)-2,2-Dioxido-4- isopropyl -1,2,3-oxathiazolidine, N-BOC protected
(4S)-2,2-Dioxido-4- isopropyl -1,2,3-oxathiazolidine, N-BOC protected
Formula:
C₁₀H₁₉NO₅S MW: 265,327 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD17018789 CAS Number: 1206227-46-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...