4-Bromo-5-chloro-1H-benzimidazole 95%
Supplier: Apollo Scientific
|
Warning
|
OR400381-1G
OR400381-250MG
APOSOR400381-1GEA
956
EUR
APOSOR400381-1G
APOSOR400381-250MG
4-Bromo-5-chloro-1H-benzimidazole 95%
4-Bromo-5-chloro-1H-benzimidazole
Formula:
C₇H₄BrClN₂ MW: 231,477 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22558142 CAS Number: 1360902-51-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...