4-Cyano-2-methoxybenzoic acid
Supplier: Apollo Scientific
|
Warning
|
OR400419-1G
APOSOR400419-1GEA
102
EUR
APOSOR400419-1G
4-Cyano-2-methoxybenzoic acid
4-Cyano-2-methoxybenzoic acid
Formula:
C₉H₇NO₃ MW: 177,157 g/mol Melting Pt: 178…180 °C Storage Temperature: Ambient |
MDL Number:
MFCD11520909 CAS Number: 89469-52-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...