7-Chloro-2-oxo-2,3-dihydro-1,3-benzoxazole

Supplier: Apollo Scientific
Warning

OR400450-1G OR400450-250MG
APOSOR400450-1GEA 303 EUR
APOSOR400450-1G APOSOR400450-250MG
7-Chloro-2-oxo-2,3-dihydro-1,3-benzoxazole
7-Chloro-2-oxo-2,3-dihydro-1,3-benzoxazole
Formula: C₇H₄ClNO₂
MW: 169,565 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11845141
CAS Number: 83972-30-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR