6-Amino-3-bromo-2-chlorobenzoic acid 97%
Supplier: Apollo Scientific
|
Warning
|
OR400466-250MG
OR400466-1G
APOSOR400466-250MGEA
58
EUR
APOSOR400466-250MG
APOSOR400466-1G
6-Amino-3-bromo-2-chlorobenzoic acid 97%
6-Amino-3-bromo-2-chlorobenzoic acid
Formula:
C₇H₅BrClNO₂ MW: 250,477 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06409110 CAS Number: 3030-19-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...