Methyl 3-amino-7-bromobenzo[b]thiophene-2-carboxylate
Supplier: Apollo Scientific
OR400500-1G
OR400500-250MG
APOSOR400500-1GEA
364
EUR
APOSOR400500-1G
APOSOR400500-250MG
Methyl 3-amino-7-bromobenzo[b]thiophene-2-carboxylate
Methyl 3-amino-7-bromobenzo[b]thiophene-2-carboxylate
Formula:
C₁₀H₈BrNO₂S MW: 286,145 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28348425 CAS Number: 1236150-66-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...