5-Bromo-2-iodo-4-methoxyaniline 97%

Supplier: Apollo Scientific
Danger

OR400545-1G OR400545-250MG
APOSOR400545-1GEA 567 EUR
APOSOR400545-1G APOSOR400545-250MG
5-Bromo-2-iodo-4-methoxyaniline 97%
5-Bromo-2-iodo-4-methoxyaniline
Formula: C₇H₇BrINO
MW: 327,945 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28348436
CAS Number: 1361021-26-2
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR