2-Amino-4-bromo-6-iodophenol, tech

Supplier: Apollo Scientific
Warning

OR400598-1G OR400598-250MG OR400598-5G
APOSOR400598-1GEA 143 EUR
APOSOR400598-1G APOSOR400598-250MG APOSOR400598-5G
2-Amino-4-bromo-6-iodophenol, tech
2-Amino-4-bromo-6-iodophenol
Formula: C₆H₅BrINO
MW: 313,918 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28397815
CAS Number: 855836-12-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR