3-Bromo-5-nitrobenzenesulphonyl chloride 95%
Supplier: Apollo Scientific
OR400678-1G
OR400678-250MG
OR400678-5G
APOSOR400678-1GEA
473
EUR
APOSOR400678-1G
APOSOR400678-250MG
APOSOR400678-5G
3-Bromo-5-nitrobenzenesulphonyl chloride 95%
3-Bromo-5-nitrobenzenesulphonyl chloride
Formula:
C₆H₃BrClNO₄S MW: 300,514 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16679614 CAS Number: 1211517-87-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...