7-Bromo-5-iodo-1,3-benzoxazole

Supplier: Apollo Scientific
Warning

OR400680-100MG OR400680-1G
APOSOR400680-100MGEA 129 EUR
APOSOR400680-100MG APOSOR400680-1G
7-Bromo-5-iodo-1,3-benzoxazole
7-Bromo-5-iodo-1,3-benzoxazole
Formula: C₇H₃BrINO
MW: 323,913 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28515360
CAS Number: 1845689-86-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR