4-Bromo-2-nitrobenzenesulphonyl chloride
Supplier: Apollo Scientific
|
Danger
|
OR400725-1G
OR400725-5G
APOSOR400725-1GEA
226
EUR
APOSOR400725-1G
APOSOR400725-5G
4-Bromo-2-nitrobenzenesulphonyl chloride
4-Bromo-2-nitrobenzenesulphonyl chloride
Formula:
C₆H₃BrClNO₄S MW: 300,514 g/mol Melting Pt: 95…97 °C Storage Temperature: Ambient |
MDL Number:
MFCD12068352 CAS Number: 89465-98-5 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...