2-Bromo-6-nitrobenzenesulphonyl chloride

Supplier: Apollo Scientific
Danger

OR400739-1G OR400739-250MG
APOSOR400739-1GEA 343 EUR
APOSOR400739-1G APOSOR400739-250MG
2-Bromo-6-nitrobenzenesulphonyl chloride
2-Bromo-6-nitrobenzenesulphonyl chloride
Formula: C₆H₃BrClNO₄S
Storage Temperature: Ambient
MDL Number: MFCD18392271
CAS Number: 1261675-40-4
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR