2-Bromo-3-iodonitrobenzene

Supplier: Apollo Scientific
Warning

OR400748-1G OR400748-5G
APOSOR400748-1GEA 352 EUR
APOSOR400748-1G APOSOR400748-5G
2-Bromo-3-iodonitrobenzene
2-Bromo-3-iodonitrobenzene
Formula: C₆H₃BrINO₂
MW: 327,902 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18391756
CAS Number: 32337-95-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR