Methyl 2-methoxy-3-methylbenzoate 95%
Supplier: Apollo Scientific
|
Warning
|
OR400880-1G
OR400880-5G
OR400880-10G
APOSOR400880-1GEA
14.7
EUR
APOSOR400880-1G
APOSOR400880-5G
APOSOR400880-10G
Methyl 2-methoxy-3-methylbenzoate 95%
Methyl 2-methoxy-3-methylbenzoate
Formula:
C₁₀H₁₂O₃ MW: 180,2 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12025375 CAS Number: 52239-62-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...