5-Bromo-3-chloro-2-methoxyaniline 95%

Supplier: Apollo Scientific
Danger

OR400940-1G OR400940-250MG OR400940-5G
APOSOR400940-1GEA 464 EUR
APOSOR400940-1G APOSOR400940-250MG APOSOR400940-5G
5-Bromo-3-chloro-2-methoxyaniline 95%
5-Bromo-3-chloro-2-methoxyaniline
Formula: C₇H₇BrClNO
Storage Temperature: Ambient
MDL Number: MFCD31561243
CAS Number: 1547274-38-3
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR