4-Iodo-2-methoxybenzyl alcohol

Supplier: Apollo Scientific
Danger

OR401048-1G OR401048-250MG
APOSOR401048-1GEA 58 EUR
APOSOR401048-1G APOSOR401048-250MG
4-Iodo-2-methoxybenzyl alcohol
4-Iodo-2-methoxybenzyl alcohol
Formula: C₈H₉IO₂
MW: 264,06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11044836
CAS Number: 210037-23-3
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR