3-Iodo-5-nitrobenzaldehyde 95%
Supplier: Apollo Scientific
|
Warning
|
OR401061-1G
OR401061-250MG
OR401061-5G
APOSOR401061-1GEA
87
EUR
APOSOR401061-1G
APOSOR401061-250MG
APOSOR401061-5G
3-Iodo-5-nitrobenzaldehyde 95%
3-Iodo-5-nitrobenzaldehyde
Formula:
C₇H₄INO₃ Storage Temperature: Ambient |
MDL Number:
MFCD18397941 CAS Number: 1261752-40-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...