6-Amino-2,3-dimethylpyridine
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-Amino-5,6-dimethylpyridine, 6-Amino-2,3-lutidine, 5,6-Dimethylpyridin-2-amine
OR40193-1G
OR40193-250MG
APOSOR40193-1GEA
39.3
EUR
APOSOR40193-1G
APOSOR40193-250MG
6-Amino-2,3-dimethylpyridine
6-Amino-2,3-dimethylpyridine
Formula:
C₇H₁₀N₂ MW: 122,168 g/mol Melting Pt: 41…43 °C Storage Temperature: Ambient |
MDL Number:
MFCD06200777 CAS Number: 57963-08-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...