1H-Indazole-7-carboxaldehyde 95%

Supplier: Apollo Scientific
Warning

Synonyms: 1H-Indazole-7-carbaldehyde, 7-Formyl-1H-indazole

OR40230-1G OR40230-250MG
APOSOR40230-1GEA 149 EUR
APOSOR40230-1G APOSOR40230-250MG
1H-Indazole-7-carboxaldehyde 95%
1H-Indazole-7-carboxaldehyde
Formula: C₈H₆N₂O
MW: 146,146 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06738281
CAS Number: 312746-72-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR