2-Hydroxypyridine-4-boronic acid 98%

Supplier: Apollo Scientific
Warning

OR40293-1G OR40293-250MG
APOSOR40293-1GEA 347 EUR
APOSOR40293-1G APOSOR40293-250MG
2-Hydroxypyridine-4-boronic acid 98%
2-Hydroxypyridine-4-boronic acid
Formula: C₅H₆BNO₃
Storage Temperature: Freezer
MDL Number: MFCD11223187
CAS Number: 902148-83-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR