3-Amino-4-isobutoxybenzoic acid
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
3-Amino-4-(2-methylpropoxy)benzoic acid, 5-Carboxy-2-isobutoxyaniline
OR40321-250MG
OR40321-500MG
APOSOR40321-250MGEA
132
EUR
APOSOR40321-250MG
APOSOR40321-500MG
3-Amino-4-isobutoxybenzoic acid
3-Amino-4-isobutoxybenzoic acid
Formula:
C₁₁H₁₅NO₃ MW: 209,242 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD12168706 CAS Number: 1096806-43-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...