4-(5-Methyl-1,3,4-oxadiazol-2-yl)benzyl alcohol

Supplier: Apollo Scientific
Warning

Synonyms: [4-(5-Methyl-1,3,4-oxadiazol-2-yl)phenyl]methanol, 2-[4-(Hydroxymethyl)phenyl]-5-methyl-1,3,4-oxadiazole

OR40512-1G OR40512-250MG
APOSOR40512-1GEA 650 EUR
APOSOR40512-1G APOSOR40512-250MG
4-(5-Methyl-1,3,4-oxadiazol-2-yl)benzyl alcohol
4-(5-Methyl-1,3,4-oxadiazol-2-yl)benzyl alcohol
Formula: C₁₀H₁₀N₂O₂
MW: 190,199 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12022274
CAS Number: 179055-98-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR