2-Bromo-7-chlorophenazine 95%

Supplier: Apollo Scientific
Warning

OR40772-1G OR40772-250MG
APOSOR40772-1GEA 111 EUR
APOSOR40772-1G APOSOR40772-250MG
2-Bromo-7-chlorophenazine 95%
2-Bromo-7-chlorophenazine
Formula: C₁₂H₆BrClN₂
MW: 293.546 g/mol
Melting Pt: 254 °C
Storage Temperature: Ambient
MDL Number: MFCD31920065
CAS Number: 29453-79-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR