2-Amino-7-chlorophenazine 95%
Supplier: Apollo Scientific
|
Warning
|
OR40773-1G
OR40773-250MG
APOSOR40773-1GEA
186
EUR
APOSOR40773-1G
APOSOR40773-250MG
2-Amino-7-chlorophenazine 95%
2-Amino-7-chlorophenazine
Formula:
C₁₂H₈ClN₃ Melting Pt: 251…253 °C Storage Temperature: Ambient |
MDL Number:
MFCD27947845 CAS Number: 23677-11-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...