4-Azaindole-2-carboxylic acid 98%

Supplier: Apollo Scientific

Synonyms: 1H-Pyrrolo[3,2-b]pyridine-2-carboxylic acid, 2-Carboxy-1H-pyrrolo[3,2b]pyridine

OR41130-250MG OR41130-1G
APOSOR41130-250MGEA 75.7 EUR
APOSOR41130-250MG APOSOR41130-1G
4-Azaindole-2-carboxylic acid 98%
4-Azaindole-2-carboxylic acid
Formula: C₈H₆N₂O₂
MW: 162,145 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09955613
CAS Number: 17288-35-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR