2-Bromo-4-chloro-6-cyclohexylphenol
Supplier: Apollo Scientific
|
Danger
|
OR41191-1G
OR41191-250MG
OR41191-5G
APOSOR41191-1GEA
26.1
EUR
APOSOR41191-1G
APOSOR41191-250MG
APOSOR41191-5G
2-Bromo-4-chloro-6-cyclohexylphenol
2-Bromo-4-chloro-6-cyclohexylphenol
Formula:
C₁₂H₁₄BrClO Storage Temperature: Ambient |
MDL Number:
MFCD32111676 CAS Number: 2367002-72-8 UN: 2923 ADR: 8+6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...