1-(4-Phenylbut-1-yl)piperazine 99%

Supplier: Apollo Scientific
Warning

Synonyms: 1-Phenyl-4-(piperazin-1-yl)butane

OR4171-2G OR4171-10G
APOSOR4171-2GEA 151 EUR
APOSOR4171-2G APOSOR4171-10G
1-(4-Phenylbut-1-yl)piperazine 99%
1-(4-Phenylbut-1-yl)piperazine
Formula: C₁₄H₂₂N₂
Boiling Pt: 110…112 °C (0,3 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD02093540
CAS Number: 97480-93-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR