5-Hydroxybenzo[b]furan

Supplier: Apollo Scientific
Warning

Synonyms: Benzo[b]furan-5-ol

OR42004-1G OR42004-250MG
APOSOR42004-1GEA 241 EUR
APOSOR42004-1G APOSOR42004-250MG
5-Hydroxybenzo[b]furan
5-Hydroxybenzo[b]furan
Formula: C₈H₆O₂
MW: 134.132 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10699412
CAS Number: 13196-10-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR