4-Bromo-2-(methylsulphonyl)benzonitrile
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
5-Bromo-2-cyanophenyl methyl sulphone
OR42106-1G
OR42106-250MG
OR42106-5G
APOSOR42106-1GEA
174
EUR
APOSOR42106-1G
APOSOR42106-250MG
APOSOR42106-5G
4-Bromo-2-(methylsulphonyl)benzonitrile
4-Bromo-2-(methylsulphonyl)benzonitrile
Formula:
C₈H₆BrNO₂S Melting Pt: 142…144 °C Storage Temperature: Ambient |
MDL Number:
MFCD12828670 CAS Number: 1208402-11-2 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...