3-Bromophenylacetylene
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
1-Bromo-3-ethynylbenzene
OR42232-1G
OR42232-25G
OR42232-5G
APOSOR42232-1GEA
21.9
EUR
APOSOR42232-1G
APOSOR42232-25G
APOSOR42232-5G
3-Bromophenylacetylene
3-Bromophenylacetylene
Formula:
C₈H₅Br MW: 181,029 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD03839983 CAS Number: 766-81-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...