1-Tosyl-1H-pyrrolo[2,3-b]pyridine-4-boronic acid, pinacol ester 97%

Supplier: Apollo Scientific
Warning

Synonyms: 1-[(4-Methylphenyl)sulphonyl]-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrolo[2,3-b]pyridine

OR42236-1G OR42236-250MG
APOSOR42236-1GEA 835 EUR
APOSOR42236-1G APOSOR42236-250MG
1-Tosyl-1H-pyrrolo[2,3-b]pyridine-4-boronic acid, pinacol ester 97%
1-Tosyl-1H-pyrrolo[2,3-b]pyridine-4-boronic acid, pinacol ester
Formula: C₂₀H₂₃BN₂O₄S
Storage Temperature: Ambient
MDL Number: MFCD09953957
CAS Number: 916176-50-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR