2-Amino-6-cyclopropylpyridine

Supplier: Apollo Scientific
Warning

Synonyms: 6-Cyclopropylpyridin-2-amine

OR42286-1G OR42286-250MG
APOSOR42286-1GEA 427 EUR
APOSOR42286-1G APOSOR42286-250MG
2-Amino-6-cyclopropylpyridine
2-Amino-6-cyclopropylpyridine
Formula: C₈H₁₀N₂
MW: 134,178 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12032770
CAS Number: 857292-66-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR