3-Amino-5-cyclopropylpyridine

Supplier: Apollo Scientific
Warning

Synonyms: 5-Cyclopropylpyridin-3-amine

OR42287-1G OR42287-250MG OR42287-5G
APOSOR42287-1GEA 190 EUR
APOSOR42287-1G APOSOR42287-250MG APOSOR42287-5G
3-Amino-5-cyclopropylpyridine
3-Amino-5-cyclopropylpyridine
Formula: C₈H₁₀N₂
Storage Temperature: Refrigerator
MDL Number: MFCD19543987
CAS Number: 1314353-68-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR