1,2-Dibromo-2,3-dichloropropane 98%
Supplier: Apollo Scientific
OR43547-25G
OR43547-5G
APOSOR43547-25GEA
0
EUR
APOSOR43547-25G
APOSOR43547-5G
1,2-Dibromo-2,3-dichloropropane 98%
1,2-Dibromo-2,3-dichloropropane
Formula:
C₃H₄Br₂Cl₂ MW: 270,778 g/mol Boiling Pt: 20 °C (18 mm Hg) Storage Temperature: Ambient |
MDL Number:
MFCD00018928 CAS Number: 70289-31-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...