2-Bromo-4-hydroxynicotinic acid 95%

Supplier: Apollo Scientific

Synonyms: 2-Bromo-4-hydroxypyridine-3-carboxylic acid, 2-Bromo-3-carboxy-4-hydroxypyridine

OR43570-1G OR43570-5G
APOSOR43570-1GEA 216 EUR
APOSOR43570-1G APOSOR43570-5G
2-Bromo-4-hydroxynicotinic acid 95%
2-Bromo-4-hydroxynicotinic acid
Formula: C₆H₄BrNO₃
MW: 218,005 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12026106
CAS Number: 1150561-81-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR