5-Chloropyrazolo[1,5-a]pyrimidine 97%

Supplier: Apollo Scientific

OR450003-10G OR450003-5G
APOSOR450003-10GEA 107 EUR
APOSOR450003-10G APOSOR450003-5G
5-Chloropyrazolo[1,5-a]pyrimidine 97%
5-Chloropyrazolo[1,5-a]pyrimidine
Formula: C₆H₄ClN₃
MW: 153,569 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04035683
CAS Number: 29274-24-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR