7-Bromobenzo[d]isoxazole 95%
Supplier: Apollo Scientific
|
Warning
|
OR450021-1G
OR450021-250MG
APOSOR450021-1GEA
335
EUR
APOSOR450021-1G
APOSOR450021-250MG
7-Bromobenzo[d]isoxazole 95%
7-Bromobenzo[d]isoxazole
Formula:
C₇H₄BrNO MW: 198.017 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11520894 CAS Number: 1260751-81-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...