3,4-Dichloroisothiazole-5-carboxylic acid

Supplier: Apollo Scientific
Warning

OR450031-5G OR450031-25G
APOSOR450031-5GEA 65.5 EUR
APOSOR450031-5G APOSOR450031-25G
3,4-Dichloroisothiazole-5-carboxylic acid
3,4-Dichloroisothiazole-5-carboxylic acid
Formula: C₄HCl₂NO₂S
MW: 198,027 g/mol
Melting Pt: 175…177 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03789232
CAS Number: 18480-53-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR