2-(2-Amino-4-oxo-thiazol-5-yl)acetic acid
Supplier: Apollo Scientific
OR450086-1G
OR450086-250MG
APOSOR450086-1GEA
174
EUR
APOSOR450086-1G
APOSOR450086-250MG
2-(2-Amino-4-oxo-thiazol-5-yl)acetic acid
2-(2-Amino-4-oxo-thiazol-5-yl)acetic acid
Formula:
C₅H₆N₂O₃S MW: 174,178 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00518998 CAS Number: 33176-41-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...