3-Bromo-5-chloropyridine-4-carbaldehyde 97%
Supplier: Apollo Scientific
|
Danger
|
OR45009-1G
OR45009-5G
APOSOR45009-1GEA
203
EUR
APOSOR45009-1G
APOSOR45009-5G
3-Bromo-5-chloropyridine-4-carbaldehyde 97%
3-Bromo-5-chloropyridine-4-carbaldehyde
Formula:
C₆H₃BrClNO Storage Temperature: Ambient |
MDL Number:
MFCD16606503 CAS Number: 1064678-66-5 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...