Methyl 2-amino-4,5-dihydro-4-thiazolecarboxylate

Supplier: Apollo Scientific

OR450100-1G OR450100-250MG
APOSOR450100-1GEA 429 EUR
APOSOR450100-1G APOSOR450100-250MG
Methyl 2-amino-4,5-dihydro-4-thiazolecarboxylate
Methyl 2-amino-4,5-dihydro-4-thiazolecarboxylate
Formula: C₅H₈N₂O₂S
MW: 160,194 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28652971
CAS Number: 66512-81-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR