Cyclopropanecarbonyl isocyanate

Supplier: Apollo Scientific

OR450131-1G OR450131-250MG
APOSOR450131-1GEA 429 EUR
APOSOR450131-1G APOSOR450131-250MG
Cyclopropanecarbonyl isocyanate
Cyclopropanecarbonyl isocyanate

May form small amount of sediment

Formula: C₅H₅NO₂
MW: 111,099 g/mol
Storage Temperature: Freezer
MDL Number: MFCD16300681
CAS Number: 69166-52-5
UN: 2206
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR