4-Nitrobenzoyl isocyanate

Supplier: Apollo Scientific

OR450133-1G OR450133-250MG
APOSOR450133-1GEA 509 EUR
APOSOR450133-1G APOSOR450133-250MG
4-Nitrobenzoyl isocyanate
4-Nitrobenzoyl isocyanate

May form small amount of sediment

Formula: C₈H₄N₂O₄
MW: 192,128 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00116469
CAS Number: 4461-37-4
UN: 2206
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR