3-Cyclobutylalanine 95%

Supplier: Apollo Scientific

OR451069-5G OR451069-1G
APOSOR451069-5GEA 1210 EUR
APOSOR451069-5G APOSOR451069-1G
3-Cyclobutylalanine 95%
3-Cyclobutylalanine
Formula: C₇H₁₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD09264349
CAS Number: 4426-06-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR