4-Cyclopropyl-benzoic acid

Supplier: Apollo Scientific
Warning

Synonyms: 4-Cyclopropylbenzoic acid

OR451110-1G OR451110-250MG OR451110-5G
APOSOR451110-1GEA 241 EUR
APOSOR451110-1G APOSOR451110-250MG APOSOR451110-5G
4-Cyclopropyl-benzoic acid
4-Cyclopropyl-benzoic acid
Formula: C₁₀H₁₀O₂
MW: 162.185 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01069076
CAS Number: 1798-82-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR