{3-[(Dimethylamino)methyl]phenyl}boronic acid hydrochloride

Supplier: Apollo Scientific
Warning

OR451432-1G OR451432-5G OR451432-10G
APOSOR451432-1GEA 42.2 EUR
APOSOR451432-1G APOSOR451432-5G APOSOR451432-10G
{3-[(Dimethylamino)methyl]phenyl}boronic acid hydrochloride
{3-[(Dimethylamino)methyl]phenyl}boronic acid hydrochloride
Formula: C₉H₁₄BNO₂·ClH
Storage Temperature: Refrigerator
MDL Number: MFCD20731144
CAS Number: 1485417-01-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR