3-Bromo-4-chloro-1H-pyrazole
Supplier: Apollo Scientific
|
Warning
|
OR451453-1G
OR451453-5G
APOSOR451453-1GEA
78.6
EUR
APOSOR451453-1G
APOSOR451453-5G
3-Bromo-4-chloro-1H-pyrazole
3-Bromo-4-chloro-1H-pyrazole
Formula:
C₃H₂BrClN₂ MW: 181,418 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD30079725 CAS Number: 27258-12-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...