Benzo[d]thiazole-2-carbonitrile 98%
Supplier: Apollo Scientific
|
Warning
|
OR45147-1G
OR45147-250MG
APOSOR45147-1GEA
270
EUR
APOSOR45147-1G
APOSOR45147-250MG
Benzo[d]thiazole-2-carbonitrile 98%
Benzo[d]thiazole-2-carbonitrile
Formula:
C₈H₄N₂S Storage Temperature: Refrigerator |
MDL Number:
MFCD08848199 CAS Number: 2602-85-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...