4-Bromophenylcyanamide
Supplier: Apollo Scientific
|
Warning
|
OR45178-1G
OR45178-250MG
APOSOR45178-1GEA
278
EUR
APOSOR45178-1G
APOSOR45178-250MG
4-Bromophenylcyanamide
4-Bromophenylcyanamide
Formula:
C₇H₅BrN₂ MW: 197.032 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD12402221 CAS Number: 60592-84-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...