N-(2-Bromophenyl)cyanamide
Supplier: Apollo Scientific
|
Warning
|
OR45180-1G
OR45180-250MG
APOSOR45180-1GEA
271
EUR
APOSOR45180-1G
APOSOR45180-250MG
N-(2-Bromophenyl)cyanamide
N-(2-Bromophenyl)cyanamide
Formula:
C₇H₅BrN₂ MW: 197,032 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD16767981 CAS Number: 343269-59-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...